Anti-PHLP1 Recombinant Antibody [Clone: Clone 25]
Catalog # PRSI24-469
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Clonality:Recombinant
- Clone:Clone 25
- Conjugation:Unconjugated
- ELISA:Yes
- Isotype:IgG kappa
- Reactivity:Timothy grass (Phleum pratense)
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:Grass Pollen Allergen
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Phl p 1, a grass pollen allerge
- Tested applications:dotblot
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: PHLP1
Clonality: Recombinant
Clone: Clone 25
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Timothy-Grass (Phleum pratense)