Order Entry
Portugal
ContactUsLinkComponent
Anti-CD4 Recombinant Antibody [Clone: 13B8.2]
Anti-CD4 Recombinant Antibody [Clone: 13B8.2]
Catalog # PRSI24-271
Supplier:  ProSci Inc.
Anti-CD4 Recombinant Antibody [Clone: 13B8.2]
Catalog # PRSI24-271
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HGNC:HGNC:1678
  • Antigen symbol:
    CD4
  • Clonality:
    Recombinant
  • Clone:
    13B8.2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    920
  • Antigen synonyms:
    Leu3/T4
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Human HSB2-T cell
  • Tested applications:
    EIA
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: HGNC:HGNC:1678
Clonality: Recombinant
Clone: 13B8.2
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human