Order Entry
Portugal
ContactUsLinkComponent
Idarubicin hydrochloride ≥98% (by HPLC)
Idarubicin hydrochloride ≥98% (by HPLC)
Catalog # ENZOALX380260M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Idarubicin hydrochloride ≥98% (by HPLC)
Catalog # ENZOALX380260M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :497.5 · 36.5 g/mol
Storage Temperature:Freezer
CAS Number:57852-57-0
Formula:C₂₆H₂₇NO₉·HCl
MDL Number:MFCD00897212