Rosiglitazone ≥97% (by HPLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:122320-73-4
:
Formula:C₁₈H₁₉N₃O₃S
MDL Number:MFCD00871760
:357.43 g/mol
Merck Index:14,08265
Storage Temperature:Refrigerator
Specification Test Results
- Purity:≥97% by HPLC
Specification Test Results
Frequently Bought Together