Order Entry
Portugal
ContactUsLinkComponent
JK 184 ≥99% (by HPLC)
JK 184 ≥99% (by HPLC)
Catalog # ENZOENZCHM1290005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
JK 184 ≥99% (by HPLC)
Catalog # ENZOENZCHM1290005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :350.4374 g/mol
CAS Number:315703-52-7
Formula:C₁₉H₁₈N₄OS
MDL Number:MFCD01044465