Idebenone ≥98% (by HPLC)
New Product
: 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methyl-1,4-benzoquinone, 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
:58186-27-9
:
:C₁₉H₃₀O₅
:MFCD00274552
:338.44 g/mol
:Ambient
- Environmentally Preferable:
- Purity:≥98% (TLC)
- Appearance:Orange solid
Frequently Bought Together