Chloramphenicol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-75-7
:
Formula:Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MDL Number:MFCD00078159
:323.13 g/mol
Merck Index:13,02087
Boiling Pt:645 °C (1013 hPa)
Melting Pt:149…153 °C
Density:1,49 g/cm³ (20 °C)
EINECS:200-287-4
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellowish-white to greyish-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:149 to 153 °C
- Loss on drying:≤0.5 % (105 °C)
- UV spectrophotometry:98.0 to 102.0 % (on dried substance)
- Specific optical rotation:+18.5° to +20.5° (20 °C, 589 nm) (c=6, ethanol)
Specification Test Results
Frequently Bought Together