β-Alanine, Sigma-Aldrich®
Catalog # SIAL05159-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:89.09 g/mol
Merck Index:13,00204
Boiling Pt:201 °C (1013 hPa)
Melting Pt:94 °C
Density:1.44 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:203-536-5
CAS Number:107-95-9
Formula:C₃H₇NO₂
MDL Number:MFCD00008200