o-Tolidine 95% pract.
:119-93-7
:
:[-C₆H₃(CH₃)-₄-NH₂]₂
:MFCD00014773
:212.29 g/mol
:12,09651
:∼361 °C (1013 hPa)
:125…132 °C
:~ 1,234 g/cm³ (20 °C)
:244 °C
:Ambient
:204-358-0
:3077
:9,III
:GHS07
:GHS08
:GHS09
- Appearance:Beige to grey to brown Powder
- Infrared spectrum:Conforms
- Melting point:125 to 132 °C
- Titration with HClO4:≥94 %
Frequently Bought Together