Order Entry
Portugal
ContactUsLinkComponent
 
Clobetasone butyrate ≥98%
Catalog # C5548-100MG
CAS Number:  
Clobetasone butyrate ≥98%
Catalog # C5548-100MG
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :478.98 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Room Temperature
CAS Number:25122-57-0
Formula:C₂₆H₃₂ClFO₅
MDL Number:MFCD00133196