3-Pyridylmethanol ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 100-55-0 Molecular Formula: C6H7NO Formula Weight: 109.13 Storage Temperature: 2°C to 8°C Flash Point: 149°(300°F) Refractive Index: 1.5450 Melting Point: -8 to -6° Boiling Point: 154°/28mm Beilstein Reference: 107851 Merck Reference: 14,6527 MDL No.: MFCD00006407
Synonyms: 3-(Hydroxymethyl)pyridine, 3-Pyridinemethanol, 3-Pyridylcarbinol, Pyridine-3-methanol, 3-Pyridylmethanol
- CAS Number:100-55-0
- Formula:C₆H₇NO
- MDL Number:MFCD00006407
- Molecular Weight:109.13 g/mol
- Merck Index:14,6527
- Boiling Pt:154 °C (28 mmHg)
- Melting Pt:–8…–6 °C
- Melting Pt:–8 to –6 °C
- Density:1.130
- Flash Pt:149 °C (300 °F)
- EINECS:202-864-6
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 100-55-0 Molecular Formula: C6H7NO Formula Weight: 109.13 Storage Temperature: 2°C to 8°C Flash Point: 149°(300°F) Refractive Index: 1.5450 Melting Point: -8 to -6° Boiling Point: 154°/28mm Beilstein Reference: 107851 Merck Reference: 14,6527 MDL No.: MFCD00006407
Synonyms: 3-(Hydroxymethyl)pyridine, 3-Pyridinemethanol, 3-Pyridylcarbinol, Pyridine-3-methanol, 3-Pyridylmethanol
- CAS Number:100-55-0
- Formula:C₆H₇NO
- MDL Number:MFCD00006407
- Molecular Weight:109.13 g/mol
- Merck Index:14,6527
- Boiling Pt:154 °C (28 mmHg)
- Melting Pt:–8…–6 °C
- Melting Pt:–8 to –6 °C
- Density:1.130
- Flash Pt:149 °C (300 °F)
- EINECS:202-864-6



