Order Entry
Portugal
ContactUsLinkComponent
4-Benzyloxyphenol 98%
Irritant
4-Benzyloxyphenol 98%
Certificates
undefined
Irritant
4-Benzyloxyphenol 98%

Specification Test Results

  • Appearance:
    Off-white to beige to brown Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    117 to 122 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together