Order Entry
Portugal
ContactUsLinkComponent
Trypan blue, Ultra Pure Grade live cell labeling
Trypan blue, Ultra Pure Grade live cell labeling
Catalog # AATB2452
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Trypan blue, Ultra Pure Grade live cell labeling
Catalog # AATB2452
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :960.82 g/mol
ADR:5,II
 :GHS07
CAS Number:72-57-1
Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
MDL Number:MFCD00003969