Cell Navigator® Cell Plasma Membrane Staining Kit *Orange Fluorescence*
AATB22680
:
- Pk:500 Tests
- Description:Cell Navigator™ cell plasma membrane staining kit *orange fluorescence*
The Cell Navigator® Plasma membrane stain kit provides a fast and uniform labelling of the plasma membrane without the cell-type differences exhibited by lectins.
- Signal persists through fixation
- Stable labeling regardless of cell type
- Multiple applications
: For research use only.