Order Entry
Portugal
ContactUsLinkComponent
Arsenic, powder, Sigma-Aldrich®
Arsenic, powder, Sigma-Aldrich®
Catalog # SIAL267961-10G
Supplier:  Merck
CAS Number:  
Arsenic, powder, Sigma-Aldrich®
Catalog # SIAL267961-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :74.92 g/mol
Density:5.7 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1558
ADR:6.1,II
 :M-Clarity™ quality level = MQ100
EINECS:231-148-6
 :GHS06
 :GHS09
CAS Number:7440-38-2
Formula:As
MDL Number:MFCD00085309