Order Entry
Portugal
ContactUsLinkComponent
Nuclear Orange™ DCS1
Nuclear Orange™ DCS1
Catalog # AATB17551
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Nuclear Orange™ DCS1
Catalog # AATB17551
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    0,5 mL
  • Description:
    Nuclear Orange™ DCS1
  • Purity:
    90%

Specifications

About this item

Our Nuclear Orange™ DCS1 is a fluorogenic, DNA-selective and cell-impermeant dye for analyzing DNA content in dead, fixed or apoptotic cells.

  • Cell-Impermeable
  • Useful For Multicolor Imaging
  • Bright Fluorescence

5 mM DMSO Solution

Caution: For research use only.