AERO CHROME* Crew Performance Surgical Gowns
Supplier: Halyard
Certificates
About this item
AERO CHROME* CREW Breathable surgical gowns combine unprecedented softness and breathability with maximum protection against fluids and pathogens.
- Meets AAMI Level 4 standard, providing the highest level of both fluid and microbial protection in the critical zones
- Proprietary fabric technology allows moisture vapor to escape while maintaining a high performance barrier
- Designed to keep your OR staff both cool and protected during long, fluid-intensive procedures
- Highest rating for resistance to ignition from surgical lasers
- Excellent abrasion and lint resistance
Certifications: Meets ASTM F1671 in the critical zones (ties, fabric, sleeve seams), per AAMI PB70:2012 Level 4, Meets AAMI Level 4, AAMI PB70:2012, EN13795 High Performance, ASTM F1671, MOCON and cup crush testing, ISO 22610 (wet),ISO 22612 (dry), ASTM F1671 and ASTM F1670, Hydrohead-20811, Per ISO 11810, Per EN Resistance to Linting and Particulates test data. Abrasion resistant per Martindale test method. Meets ASTM D4966.
Specifications
- Catalog No:
- HALY48399
- HALY48401
- HALY48400
- HALY48392
- HALY48394
- HALY48393
- Closure type:
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long