Order Entry
Portugal
ContactUsLinkComponent
Recombinant Human IL1A (from HEK293 cells)
Recombinant Human IL1A (from HEK293 cells)
Catalog # ABNOP8207.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Human IL1A (from HEK293 cells)
Catalog # ABNOP8207.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Gene ID:
    P01583
  • Protein/peptide name:
    IL1A

Specifications

About this item