Recombinant Human BTC (from E. coli)
Catalog # ABNOP7462.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:P35070
- Protein/peptide name:BTC
- Sequence:32a.a.-111a.a.