Anti-PTEN (pSer380) Rabbit Monoclonal Antibody [clone: NA9] (PE)
Catalog # ABNOMAB23468.100
Supplier: Abnova
Specifications
- Antigen symbol:PTEN
- Clonality:Monoclonal
- Clone:NA9
- Conjugation:PE (Phycoerythrin)
- Host:Rabbit
- Reactivity:Human
- Modification:pSer380
- Immunogen:Rabbit monoclonal antibody raised against synthetic phosphopeptide of human PTEN
- Pk:100 AU