Anti-MAP2K4 (pSer257) Rabbit Monoclonal Antibody [clone: C5] (APC)
Catalog # ABNOMAB23474.100
Supplier: Abnova
Specifications
- Antigen symbol:MAP2K4
- Clonality:Monoclonal
- Clone:C5
- Conjugation:APC (Allophycocyanin)
- Host:Rabbit
- Reactivity:Human
- Modification:pSer257
- Immunogen:Rabbit monoclonal antibody raised against synthetic phosphopeptide of human MAP2K4
- Pk:100 AU