Order Entry
Portugal
ContactUsLinkComponent
Anti-PRKCA Rabbit Monoclonal Antibody [clone: F1] (APC)
Anti-PRKCA Rabbit Monoclonal Antibody [clone: F1] (APC)
Catalog # ABNOMAB23495.100
Supplier:  Abnova
Anti-PRKCA Rabbit Monoclonal Antibody [clone: F1] (APC)
Catalog # ABNOMAB23495.100
Supplier:  Abnova

Specifications

  • Antigen symbol:
    PRKCA
  • Clonality:
    Monoclonal
  • Clone:
    F1
  • Conjugation:
    APC (Allophycocyanin)
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Modification:
    pThr497
  • Immunogen:
    Rabbit monoclonal antibody raised against synthetic phosphopeptide of human PRKCA
  • Pk:
    100 AU

Specifications

About this item