Anti-JAK1 (pTyr1022/1023) Rabbit Monoclonal Antibody [clone: F11] (PE)
Catalog # ABNOMAB23535.100
Supplier: Abnova
Specifications
- Antigen symbol:JAK1
- Clonality:Monoclonal
- Clone:F11
- Conjugation:PE (Phycoerythrin)
- Host:Rabbit
- Reactivity:Human
- Modification:pTyr1022/1023
- Immunogen:Rabbit monoclonal antibody raised against synthetic phosphopeptide of human JAK1.
- Pk:100 AU