Anti-FOS (pSer32) Rabbit Monoclonal Antibody [clone: BA9] (PE)
Catalog # ABNOMAB23543.100
Supplier: Abnova
Specifications
- Antigen symbol:FOS
- Clonality:Monoclonal
- Clone:BA9
- Conjugation:PE (Phycoerythrin)
- Host:Rabbit
- Reactivity:Human
- Modification:pSer32
- Immunogen:Rabbit monoclonal antibody raised against synthetic phosphopeptide of human FOS.
- Pk:100 AU