Crime scene V: The abandoned shed lab activity
761-0309
- Description:Crime Scene: The Abandoned Shed, laboratory activity set
- Pk:1
Become the crime scene investigator charged with conducting five blood analysis tests to help solve the mystery of the abandoned shed, which seems to be the centre of a lot more than garden tool storage. In this activity police are notified by a mysterious caller with information that has led them to the scene of a horrible crime of murder. To solve the case student conduct ABO and Rh blood typing, Kastle-Meyer, Benzidine, and Barr Body tests to identify the killer.
- Blood evidence analysis solves the crime
- Multiple techniques
: Set includes materials for three classes of 24 students along with instructions and reference materials in a bound booklet for easy duplication.