Order Entry
Portugal
ContactUsLinkComponent
Stainless Steel Shovels
Stainless Steel Shovels
Catalog # SAMPA246-10
Supplier:  Sampling Systems
undefined
Stainless Steel Shovels
Catalog # SAMPA246-10
Supplier:  Sampling Systems
Specifications
  • Description:
    Stainless steel shovel
  • Length:
    1155 mm
  • Pk:
    1
Specifications
About this item
This stainless shovel is perfect for all general purpose work.
  • Made from chemically resistant 316L stainless steel
  • Ergonomic handle
  • Highly polished finish for easy cleaning
Made from high quality corrosion-resistant 316L stainles steel, it is crevice-free and has a high polish finish so can be easily cleaned once used.