Order Entry
Portugal
ContactUsLinkComponent
Culti-Loops™ Papiliotrema laurentii ATCC™ 66036™
Culti-Loops™ Papiliotrema laurentii ATCC™ 66036™
Catalog # OXOIR4601711
CAS Number:  
undefined
Culti-Loops™ Papiliotrema laurentii ATCC™ 66036™
Catalog # OXOIR4601711
CAS Number:  
Specifications
  • Pk:
    5
  • Description:
    Papiliotrema laurentii ATCC™ 66036™
  • Environmentally Preferable:
Specifications
About this item
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.