Calcium hydroxide ≥95.0%, powder FCC
Catalog # SPCMC1116-2.5KG
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Poly pail
- Pk:2,5 kg
- Seq:4076
Specifications
Specification Test Results
- Assay [Ca(OH)₂]:95.0 - 100.5%
- Acid-Insoluble Substances:max. 0.5 %
- Arsenic (As):max. 3 mg/kg
- Carbonate:To Pass Test
- Fluoride:max. 0.005 %
Specification Test Results
About this item
:
:74.09 g/mol
Storage Temperature:Ambient
UN:3262
ADR:8,III
:GHS05
:GHS07
CAS Number:1305-62-0
Formula:Ca(OH)₂
MDL Number:MFCD00010901