Anti-Bcl-2 Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)
BOSSBS-15533R-A350
: Bioss
- Antibody type:Primary
- Antigen name:Bcl-2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:51-150/239
- Form:Liquid
- Gene ID:596
- Antigen synonyms:PPP1R50|BCL2|Apoptosis regulator Bcl-2|Bcl-2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:26 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Bcl-2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Suppresses apoptosis in a variety of cell systems including factor-dependent lymphohematopoietic and neural cells. Regulates cell death by controlling the mitochondrial membrane permeability. Appears to function in a feedback loop system with caspases. Inhibits caspase activity either by preventing the release of cytochrome c from the mitochondria and/or by binding to the apoptosis-activating factor (APAF-1).
Type: Primary
Antigen: Bcl-2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 350
Public Immunogen Range: 51-150/239
Host: Rabbit
Isotype: IgG
Reactivity: Human