Order Entry
Portugal
ContactUsLinkComponent
 
ReliaPrep™ RNA Tissue Miniprep System
 :  Promega
 
undefined
ReliaPrep™ RNA Tissue Miniprep System
 :  Promega

 

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Storage conditions:
    15...30 °C
  • Description:
    ReliaPrep RNA tissue Miniprep system

 

Frequently Bought Together