Trospium Chloride, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
4 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1699300.
- 1699311.
- 1699322.
- 1699333.
- Description:
- Trospium Chloride
- Trospium Chloride Related Compound A
- Trospium Chloride Related Compound B
- Trospium Chloride Related Compound C
- Synonym(s):
- Benzilic Acid
- (1R,3r,5S)-8-azabicyclo[3.2.1]octan-3-yl hydroxydiphenylacetate hydrochloride
- (1R, 3r, 5S)-3-Hydroxyspiro[8-azoniabicyclo[3.2.1]octane-8, 1'-pyrrolidinium]chloride
- CAS No.:
- 10405-02-4
- 76-93-7
- 63516-30-3
- 3464-71-9
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1699300.Trospium ChlorideSpecifications:
More
Specifications
Cat. No.1699300.DescriptionTrospium ChlorideSynonym(s)CAS No.10405-02-4 - 1699311.Trospium Chloride Related Compound ATrospium Chloride Related Compound AUSP reference standardVial20 mgLog in for availability1 * 20 mg1 090,00 €Specifications:
More
Specifications
Cat. No.1699311.DescriptionTrospium Chloride Related Compound ASynonym(s)Benzilic AcidCAS No.76-93-7 - 1699322.Trospium Chloride Related Compound BTrospium Chloride Related Compound BUSP reference standardVial20 mgLog in for availability1 * 20 mg1 180,00 €Specifications:
More
Specifications
Cat. No.1699322.DescriptionTrospium Chloride Related Compound BSynonym(s)(1R,3r,5S)-8-azabicyclo[3.2.1]octan-3-yl hydroxydiphenylacetate hydrochlorideCAS No.63516-30-3 - 1699333.Trospium Chloride Related Compound CTrospium Chloride Related Compound CUSP reference standardVial20 mgLog in for availability1 * 20 mg1 190,00 €Specifications:
More
Specifications
Cat. No.1699333.DescriptionTrospium Chloride Related Compound CSynonym(s)(1R, 3r, 5S)-3-Hydroxyspiro[8-azoniabicyclo[3.2.1]octane-8, 1'-pyrrolidinium]chlorideCAS No.3464-71-9
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



