Chloramphenicol, powder
Catalog # MDTC61-239-RI
Supplier: Corning
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:323.13 g/mol
Storage Temperature:Ambient
CAS Number:56-75-7
Formula:Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MDL Number:MFCD00078159