Order Entry
Portugal
ContactUsLinkComponent
Y-connectors, b.safe
Y-connectors, b.safe
Catalog # BOHLR1220-03
Supplier:  Bohlender
undefined
Y-connectors, b.safe
Catalog # BOHLR1220-03
Supplier:  Bohlender
Specifications
  • Hole Ø:
    14,5 mm
  • Thread:
    3× GL 25 (male)
  • Description:
    Y-connectors
  • Material:
    PE
  • Pk:
    1
Specifications
About this item
Combine 2 b.safe Waste Tubes to one outlet.
  • Made of static dissipative PE-EX
  • With optional wall mount
Three connections male thread GL 25. Two bores with Ø 5,5 mm for optional wall mount.