Tubing rolls, PFA, BOLA
Non-porous, transparent, gastight tubing.
- Most universal chemical resistance
- Free from extractable agents
- Physiologically safe
- Non-adhesive surface
- Very good sliding and dielectric characteristics
- Oxygen value more than 95
- Resistant to irradiation and weather
- Mechanical strength even at high temperatures
Perfect tubing for aggressive and pure liquids or gases. Can be sterilized in autoclaves.
: Flame retardant according to UL94V0.
Frequently Bought Together