CXR reference dye
Catalog # PROMC5411
Supplier: Promega
CAS Number:
Specifications
- Pk:100 µl
- Description:CXR Reference Dye
Specifications
About this item
Use Carboxy-X-Rhodamine (CXR) Reference Dye (identical to ROX™ dye) with instruments that require a higher level of reference dye that than in the GoTaq® qPCR Master Mix. Component of GoTaq® qPCR and RT-qPCR Systems.
- Use with instruments that require a higher level of reference dye
- Component of GoTaq® qPCR and RT-qPCR Systems
- Excitation at 580nm and emission at 602nm
Caution: Storage temperature: -30°C to -10°C