Order Entry
Portugal
ContactUsLinkComponent
Sheep recombinant GH A
Sheep recombinant GH A
Catalog # APOSBITP1081-1MG
Supplier:  Apollo Scientific
CAS Number:  
Sheep recombinant GH A
Catalog # APOSBITP1081-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Sheep
  • Protein synonyms:
    Growth Hormone Antagonist
  • Protein/peptide name:
    GH A

Specifications

About this item