2-(Trifluoromethoxy)terephthalic acid 95+%
Catalog # APOSPC7439TK-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:250.13 g/mol
Melting Pt:270…272 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
CAS Number:175278-21-4
Formula:C₉H₅F₃O₅
MDL Number:MFCD00203986