
Silver(I) hexafluorophosphate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:26042-63-7
:
Formula:AgF₆P
MDL Number:MFCD00003415
:252.83
Melting Pt:102 °C (dec.)
Storage Temperature:Ambient
EINECS:247-428-6
UN:3260
ADR:8,II
:GHS05
Frequently Bought Together