N,N-Diisopropylethylamine trihydrofluoride
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: N-Ethyl-N-isopropylpropan-2-amine trihydrofluoride
CAS Number:131600-43-6
:
Formula:C₈H₂₂F₃N
MDL Number:MFCD00144880
:189.266 g/mol
Boiling Pt:93…95 °C/4mmHg
Density:0.965 g/cm³ (25 °C)
Flash Pt:40.5 °C
UN:3286
ADR:3+6.1+8,II
:GHS02
:GHS05
:GHS06
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together