

Ethyl-5H-octafluorovalerate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2795-50-8
:
Formula:C₇H₆F₈O₂
MDL Number:MFCD00153153
:274.11 g/mol
Boiling Pt:180 °C
Density:1.453 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3272
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together