
[5-(Trifluoromethyl)-2-pyridinyl]methanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:31181-84-7
:
Formula:C₇H₆F₃NO
MDL Number:MFCD04117762
:177.13 g/mol
Boiling Pt:76…78 °C/1mmHg
:GHS07
Storage Temperature:Ambient
Frequently Bought Together