2-((tert-Butoxycarbonyl)amino)-2-cyclobutylacetic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:811460-95-4
:
Formula:C₁₁H₁₉NO₄
MDL Number:MFCD09264345
:229.27 g/mol
Storage Temperature:Ambient
Frequently Bought Together