Order Entry
Portugal
ContactUsLinkComponent
3-Chlorobutanone 96%
3-Chlorobutanone 96%
Catalog # APOSOR921276-1KG
Supplier:  Apollo Scientific
CAS Number:  
3-Chlorobutanone 96%
Catalog # APOSOR921276-1KG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :106.55 g/mol
Storage Temperature:Ambient
CAS Number:4091-39-8
Formula:C₄H₇ClO
MDL Number:MFCD00000865