Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Portugal
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(2-Phenyl-1H-imidazol-4-yl)methanol
(2-Phenyl-1H-imidazol-4-yl)methanol
(2-Phenyl-1H-imidazol-4-yl)methanol
APOSOR8526-1G
:
Apollo Scientific
:
(2-Phenyl-1H-imidazol-4-yl)methanol
APOSOR8526-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
144…146 °C
:
174.2 g/mol
:
Ambient
:
43002-54-6
:
C₁₀H₁₀N₂O
:
MFCD06202903