3-Bromo-4-methoxybiphenyl
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:74447-73-7
:
Formula:C₁₃H₁₁BrO
MDL Number:MFCD00079720
:263.14 g/mol
Melting Pt:73…80 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together