
2-Bromo-6-nitrobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:38876-67-4
:
Formula:C₇H₄BrNO₄
MDL Number:MFCD13688997
:246.0200 g/mol
Melting Pt:177…178 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together