
{2-Methylimidazo[1,2-a]pyridin-3-yl}methanol 90+%
Catalog # APOSOR4417-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:162.19 g/mol
Melting Pt:157…158 °C
:GHS07
Storage Temperature:Ambient
CAS Number:30489-44-2
Formula:C₉H₁₀N₂O
MDL Number:MFCD07368506