4'-(Methylthio)acetophenone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1778-09-2
:
Formula:C₉H₁₀OS
MDL Number:MFCD00039835
:166.24 g/mol
Boiling Pt:135 °C/1mmHg
Melting Pt:80…82 °C
Storage Temperature:Ambient
EINECS:217-213-1
:GHS07
Frequently Bought Together