3,5-Dichlorobenzylamine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:39989-43-0
:
Formula:C₇H₇Cl₂N
MDL Number:MFCD00052681
:176.04 g/mol
Boiling Pt:141…144 °C/15mmHg
Density:1.32 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together