Order Entry
Portugal
ContactUsLinkComponent
5-(Hydroxymethyl)-6-phenyl-4,5-dihydro-3(2H)-pyridazinone
Irritant
5-(Hydroxymethyl)-6-phenyl-4,5-dihydro-3(2H)-pyridazinone
Catalog # APOSOR12101-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-(Hydroxymethyl)-6-phenyl-4,5-dihydro-3(2H)-pyridazinone
Catalog # APOSOR12101-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:163…165 °C
 :204.23 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:23239-13-6
Formula:C₁₁H₁₂N₂O₂
MDL Number:MFCD01692709